MilliporeSigma
  • Ropivacaine Promotes Axon Regeneration by Regulating Nav1.8-mediated Macrophage Signaling after Sciatic Nerve Injury in Rats.
Ropivacaine Promotes Axon Regeneration by Regulating Nav1.8-mediated Macrophage Signaling after Sciatic Nerve Injury in Rats.
Anesthesiology (2023-09-05)
Yongchen Cui, Xiaofeng Wang, Yang Xu, Yue Cao, Gang Luo, Zhe Zhao, Junfeng Zhang
ABSTRACT
Continuous nerve block with ropivacaine is commonly performed after repair surgery for traumatic peripheral nerve injuries. After peripheral nerve injury, tetrodotoxin-resistant voltage-gated sodium channel Nav1.8 is upregulated and contributes to macrophage inflammation. This study investigated whether ropivacaine promotes peripheral nerve regeneration through Nav1.8-mediated macrophage signaling. A sciatic nerve transection-repair (SNT) model was established in adult Sprague-Dawley rats of both sexes. The rats received 0.2% ropivacaine or 10 μM Nav1.8-selective inhibitor A-803467 around the injured site or near the sacrum for 3 days. Nerve regeneration was evaluated using behavioral, electrophysiologic, and morphological examinations. Moreover, myelin debris removal, macrophage phenotype, Nav1.8 expression, and neuropeptide expression were assessed using immunostaining, enzyme-linked immunosorbent assay, and Western blotting. Compared to the SNT-plus-vehicle group, the sensory, motor, and sensory-motor coordination functions of the two ropivacaine groups were significantly improved. Electrophysiologic (mean ± SD: recovery index of amplitude, vehicle 0.43 ± 0.17 vs. ropivacaine 0.83 ± 0.25, n = 11, P < 0.001) and histological analysis collectively indicated that ropivacaine significantly promoted axonal regrowth (percentage of neurofilament 200 [NF-200]-positive area: vehicle 19.88 ± 2.81 vs. ropivacaine 31.07 ± 2.62, n = 6, P < 0.001). The authors also found that, compared to the SNT-plus-vehicle group, the SNT-plus-ropivacaine group showed faster clearance of myelin debris, accompanied by significantly increased macrophage infiltration and transition from the M1 to M2 phenotype. Moreover, ropivacaine significantly attenuated Nav1.8 upregulation at 9 days after sciatic nerve transection (vehicle 4.12 ± 0.30-fold vs. ropivacaine 2.75 ± 0.36-fold, n = 5, P < 0.001), which coincided with the increased expression of chemokine ligand 2 and substance P. Similar changes were observed when using the selective Nav1.8 channel inhibitor A-803467. Continuous nerve block with ropivacaine promotes the structural and functional recovery of injured sciatic nerves, possibly by regulating Nav1.8-mediated macrophage signaling.
Sigma-Aldrich
Monoclonal Anti-S-100 (β-Subunit) antibody produced in mouse, clone SH-B1, ascites fluid
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 129-138, clone 1, culture supernatant, clone 1, Chemicon®
Sigma-Aldrich
Anti-Neurofilament 200kDa Antibody, clone NE14, Alexa Fluor 555 Conjugate, clone NE14, from mouse, ALEXA FLUOR 555