MilliporeSigma
Remodeling oncogenic transcriptomes by small molecules targeting NONO.
Nature chemical biology (2023-03-03)
Stefan G Kathman, Seong Joo Koo, Garrett L Lindsey, Hsuan-Lin Her, Steven M Blue, Haoxin Li, Steffen Jaensch, Jarrett R Remsberg, Kay Ahn, Gene W Yeo, Brahma Ghosh, Benjamin F Cravatt
ABSTRACT
Much of the human proteome is involved in mRNA homeostasis, but most RNA-binding proteins lack chemical probes. Here we identify electrophilic small molecules that rapidly and stereoselectively decrease the expression of transcripts encoding the androgen receptor and its splice variants in prostate cancer cells. We show by chemical proteomics that the compounds engage C145 of the RNA-binding protein NONO. Broader profiling revealed that covalent NONO ligands suppress an array of cancer-relevant genes and impair cancer cell proliferation. Surprisingly, these effects were not observed in cells genetically disrupted for NONO, which were instead resistant to NONO ligands. Reintroduction of wild-type NONO, but not a C145S mutant, restored ligand sensitivity in NONO-disrupted cells. The ligands promoted NONO accumulation in nuclear foci and stabilized NONO-RNA interactions, supporting a trapping mechanism that may prevent compensatory action of paralog proteins PSPC1 and SFPQ. These findings show that NONO can be co-opted by covalent small molecules to suppress protumorigenic transcriptional networks.
Sigma-Aldrich
Monoclonal Anti-SFPQ antibody produced in mouse, clone 6D7, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-PSPC1 antibody, Mouse monoclonal, clone 1L4, purified from hybridoma cell culture