MilliporeSigma
  • A quantitative model for virus uncoating predicts influenza A infectivity.
A quantitative model for virus uncoating predicts influenza A infectivity.
Cell reports (2023-12-17)
Alina Artcibasova, Longlong Wang, Stephanie Anchisi, Yohei Yamauchi, Mirco Schmolke, Patrick Matthias, Jörg Stelling
ABSTRACT
For virus infection of new host cells, the disassembly of the protective outer protein shell (capsid) is a critical step, but the mechanisms and host-virus interactions underlying the dynamic, active, and regulated uncoating process are largely unknown. Here, we develop an experimentally supported, multiscale kinetics model that elucidates mechanisms of influenza A virus (IAV) uncoating in cells. Biophysical modeling demonstrates that interactions between capsid M1 proteins, host histone deacetylase 6 (HDAC6), and molecular motors can physically break the capsid in a tug-of-war mechanism. Biochemical analysis and biochemical-biophysical modeling identify unanchored ubiquitin chains as essential and allow robust prediction of uncoating efficiency in cells. Remarkably, the different infectivity of two clinical strains can be ascribed to a single amino acid variation in M1 that affects binding to HDAC6. By identifying crucial modules of viral infection kinetics, the mechanisms and models presented here could help formulate novel strategies for broad-range antiviral treatment.
Sigma-Aldrich
Anti-Actin-pan, C-Terminal antibody produced in rabbit, affinity isolated antibody