MilliporeSigma
  • Biochemical and physiological characterization of the pyruvate formate-lyase Pfl1 of Chlamydomonas reinhardtii, a typically bacterial enzyme in a eukaryotic alga.
Biochemical and physiological characterization of the pyruvate formate-lyase Pfl1 of Chlamydomonas reinhardtii, a typically bacterial enzyme in a eukaryotic alga.
Eukaryotic cell (2008-02-05)
Anja Hemschemeier, Jessica Jacobs, Thomas Happe
ABSTRACT
The unicellular green alga Chlamydomonas reinhardtii has a special type of anaerobic metabolism that is quite unusual for eukaryotes. It has two oxygen-sensitive [Fe-Fe] hydrogenases (EC 1.12.7.2) that are coupled to photosynthesis and, in addition, a formate- and ethanol-producing fermentative metabolism, which was proposed to be initiated by pyruvate formate-lyase (Pfl; EC 2.3.1.54). Pfl enzymes are commonly found in prokaryotes but only rarely in eukaryotes. Both the hydrogen- and the formate/ethanol-producing pathways are involved in a sustained anaerobic metabolism of the alga, which can be induced by sulfur depletion in illuminated cultures. Before now, the presence of a Pfl protein in C. reinhardtii was predicted from formate secretion and the homology of the deduced protein of the PFL1 gene model to known Pfl enzymes. In this study, we proved the formate-producing activity of the putative Pfl1 enzyme by heterologous expression of the C. reinhardtii PFL1 cDNA in Escherichia coli and subsequent in vitro activity tests of the purified protein. Furthermore, a Pfl-deficient E. coli strain secretes formate when expressing the PFL1 cDNA of C. reinhardtii. We also examined the Pfl1 fermentation pathway of C. reinhardtii under the physiological condition of sulfur depletion. Genetic and biochemical analyses show that sulfur-depleted algae express genes encoding enzymes acting downstream of Pfl1 and also potentially ethanol-producing enzymes, such as pyruvate decarboxylase (EC 4.1.1.1) or pyruvate ferredoxin oxidoreductase (EC 1.2.7.1). The latter enzymes might substitute for Pfl1 activity when Pfl1 is specifically inhibited by hypophosphite.