MilliporeSigma
Coenzyme Q10 supplementation - In ageing and disease.
Mechanisms of ageing and development (2021-06-16)
Jan Aaseth, Jan Alexander, Urban Alehagen
ABSTRACT
Coenzyme Q10 (CoQ10) is an essential component of the mitochondrial electron transport chain. It is also an antioxidant in cellular membranes and lipoproteins. All cells produce CoQ10 by a specialized cytoplasmatic-mitochondrial pathway. CoQ10 deficiency can result from genetic failure or ageing. Some drugs including statins, widely used by inter alia elderly, may inhibit endogenous CoQ10 synthesis. There are also chronic diseases with lower levels of CoQ10 in tissues and organs. High doses of CoQ10 may increase both circulating and intracellular levels, but there are conflicting results regarding bioavailability. Here, we review the current knowledge of CoQ10 biosynthesis and primary and acquired CoQ10 deficiency, and results from clinical trials based on CoQ10 supplementation. There are indications that supplementation positively affects mitochondrial deficiency syndrome and some of the symptoms of ageing. Cardiovascular disease and inflammation appear to be alleviated by the antioxidant effect of CoQ10. There is a need for further studies and well-designed clinical trials, with CoQ10 in a formulation of proven bioavailability, involving a greater number of participants undergoing longer treatments in order to assess the benefits of CoQ10 treatment in neurodegenerative disorders, as well as in metabolic syndrome and its complications.
Sigma-Aldrich
Coenzyme Q10, ≥98% (HPLC)