MilliporeSigma
  • RNF31 promotes proliferation and invasion of hepatocellular carcinoma via nuclear factor kappaB activation.
RNF31 promotes proliferation and invasion of hepatocellular carcinoma via nuclear factor kappaB activation.
Scientific reports (2024-01-04)
Kouki Hoshino, Seshiru Nakazawa, Takehiko Yokobori, Kei Hagiwara, Norihiro Ishii, Mariko Tsukagoshi, Takamichi Igarashi, Kenichiro Araki, Norifumi Harimoto, Fuminori Tokunaga, Ken Shirabe
ABSTRACT
RNF31 is a multifunctional RING finger protein implicated in various inflammatory diseases and cancers. It functions as a core component of the linear ubiquitin chain assembly complex (LUBAC), which activates the nuclear factor kappaB (NF-κB) pathway via the generation of the Met1-linked linear ubiquitin chain. We aimed to clarify the role of RNF31 in the pathogenesis of hepatocellular carcinoma (HCC) and its relevance as a therapeutic target. High RNF31 expression in HCC, assessed by both immunohistochemistry and mRNA levels, was related to worse survival rates among patients with HCC. In vitro experiments showed that RNF31 knockdown in HCC cell lines led to decreased cell proliferation and invasion, as well as suppression of tumor necrosis factor (TNF)-α-induced NF-κB activation. Treatment with HOIPIN-8, a specific LUBAC inhibitor that suppresses RNF31 ubiquitin ligase (E3) activity, showed similar effects, resulting in decreased cell proliferation and invasion. Our clinical and in vitro data showed that RNF31 is a prognostic factor for HCC that promotes tumor aggressiveness via NF-κB activation.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-RNF31 antibody produced in rabbit, affinity isolated antibody