MilliporeSigma
  • Rac1 mediates cadherin-11 induced cellular pathogenic processes in aortic valve calcification.
Rac1 mediates cadherin-11 induced cellular pathogenic processes in aortic valve calcification.
Cardiovascular pathology : the official journal of the Society for Cardiovascular Pathology (2022-01-26)
Kiran A Vaidya, Matthew P Donnelly, Ablajan Mahmut, Jae Woong Jang, Terence W Gee, Marine-Ayan Ibrahim Aibo, Robert Bossong, Clare Hall, Sanjay Samb, Jonathan Chen, Jonathan T Butcher
ABSTRACT
Calcific aortic valve disease (CAVD), a major cause for surgical aortic valve replacement, currently lacks available pharmacological treatments. Cadherin-11 (Cad11), a promising therapeutic target, promotes aortic valve calcification in vivo, but direct Cad11 inhibition in clinical trials has been unsuccessful. Targeting of downstream Cad11 effectors instead may be clinically useful; however, the downstream effectors that mediate Cad11-induced aortic valve cellular pathogenesis have not been investigated. Immunofluorescence of calcified human aortic valves revealed that GTP-Rac1 is highly upregulated in calcified leaflets and is 2.15 times more co-localized with Cad11 in calcified valves than GTP-RhoA. Using dominant negative mutants in porcine aortic valve interstitial cells (PAVICs), we show that Cad11 predominantly regulates Runx2 nuclear localization via Rac1. Rac1-GEF inhibition via NSC23766 effectively reduces calcification in ex vivo porcine aortic valve leaflets treated with osteogenic media by 2.8-fold and also prevents Cad11-induced cell migration, compaction, and calcification in PAVICs. GTP-Rac1 and Trio, a known Cad11 binding partner and Rac1-GEF, are significantly upregulated in Nfatc1Cre; R26-Cad11Tg/Tg (Cad11 OX) mice that conditionally overexpress Cad11 in the heart valves by 3.1-fold and 6.3-fold, respectively. Finally, we found that the Trio-specific Rac1-GEF inhibitor, ITX3, effectively prevents Cad11-induced calcification and Runx2 induction in osteogenic conditions. Here we show that Cad11 induces many cellular pathogenic processes via Rac1 and that Rac1 inhibition effectively prevents many Cad11-induced aortic disease phenotypes. These findings highlight the therapeutic potential of blocking Rac1-GEFs in CAVD.
Sigma-Aldrich
ITX3, ≥98% (HPLC)
Pricing and availability is not currently available.