MilliporeSigma
  • SARIFA as a new histopathological biomarker is associated with adverse clinicopathological characteristics, tumor-promoting fatty-acid metabolism, and might predict a metastatic pattern in pT3a prostate cancer.
SARIFA as a new histopathological biomarker is associated with adverse clinicopathological characteristics, tumor-promoting fatty-acid metabolism, and might predict a metastatic pattern in pT3a prostate cancer.
BMC cancer (2024-01-13)
Johanna S Enke, Matthias Groß, Bianca Grosser, Eva Sipos, Julie Steinestel, Phillip Löhr, Johanna Waidhauser, Constantin Lapa, Bruno Märkl, Nic G Reitsam
ABSTRACT
Recently, we introduced Stroma-AReactive-Invasion-Front-Areas (SARIFA) as a novel hematoxylin-eosin (H&E)-based histopathologic prognostic biomarker for various gastrointestinal cancers, closely related to lipid metabolism. To date, no studies on SARIFA, which is defined as direct tumor-adipocyte-interaction, beyond the alimentary tract exist. Hence, the objective of our current investigation was to study the significance of SARIFA in pT3a prostate cancer (PCa) and explore its association with lipid metabolism in PCa as lipid metabolism plays a key role in PCa development and progression. To this end, we evaluated SARIFA-status in 301 radical prostatectomy specimens and examined the relationship between SARIFA-status, clinicopathological characteristics, overall survival, and immunohistochemical expression of FABP4 and CD36 (proteins closely involved in fatty-acid metabolism). Additionally, we investigated the correlation between SARIFA and biochemical recurrence-free survival (BRFS) and PSMA-positive recurrences in PET/CT imaging in a patient subgroup. Moreover, a quantitative SARIFA cut-off was established to further understand the underlying tumor biology. SARIFA positivity occurred in 59.1% (n = 178) of pT3a PCas. Our analysis demonstrated that SARIFA positivity is strongly associated with established high-risk features, such as R1 status, extraprostatic extension, and higher initial PSA values. Additionally, we observed an upregulation of immunohistochemical CD36 expression specifically at SARIFAs (p = 0.00014). Kaplan-Meier analyses revealed a trend toward poorer outcomes, particularly in terms of BRFS (p = 0.1). More extensive tumor-adipocyte interaction, assessed as quantity-dependent SARIFA-status on H&E slides, is also significantly associated with high-risk features, such as lymph node metastasis, and seems to be associated with worse survival outcomes (p = 0.16). Moreover, SARIFA positivity appeared to be linked to more distant lymph node and bone metastasis, although statistical significance was slightly not achieved (both p > 0.05). This is the first study to introduce SARIFA as easy-and-fast-to-assess H&E-based biomarker in locally advanced PCa. SARIFA as the histopathologic correlate of a distinct tumor biology, closely related to lipid metabolism, could pave the way to a more detailed patient stratification and to the development of novel drugs targeting lipid metabolism in pT3a PCa. On the basis of this biomarker discovery study, further research efforts on the prognostic and predictive role of SARIFA in PCa can be designed.
Sigma-Aldrich
Anti-CD36 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution