MilliporeSigma
  • A stimulus-contingent positive feedback loop enables IFN-β dose-dependent activation of pro-inflammatory genes.
A stimulus-contingent positive feedback loop enables IFN-β dose-dependent activation of pro-inflammatory genes.
Molecular systems biology (2023-03-18)
Catera L Wilder, Diane Lefaudeux, Raisa Mathenge, Kensei Kishimoto, Alma Zuniga Munoz, Minh A Nguyen, Aaron S Meyer, Quen J Cheng, Alexander Hoffmann
ABSTRACT
Type I interferons (IFN) induce powerful antiviral and innate immune responses via the transcription factor, IFN-stimulated gene factor (ISGF3). However, in some pathological contexts, type I IFNs are responsible for exacerbating inflammation. Here, we show that a high dose of IFN-β also activates an inflammatory gene expression program in contrast to IFN-λ3, a type III IFN, which elicits only the common antiviral gene program. We show that the inflammatory gene program depends on a second, potentiated phase in ISGF3 activation. Iterating between mathematical modeling and experimental analysis, we show that the ISGF3 activation network may engage a positive feedback loop with its subunits IRF9 and STAT2. This network motif mediates stimulus-specific ISGF3 dynamics that are dependent on ligand, dose, and duration of exposure, and when engaged activates the inflammatory gene expression program. Our results reveal a previously underappreciated dynamical control of the JAK-STAT/IRF signaling network that may produce distinct biological responses and suggest that studies of type I IFN dysregulation, and in turn therapeutic remedies, may focus on feedback regulators within it.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Anti-IRF-9 Antibody, clone 6F1-H5, clone 6F1-H5, from mouse