MilliporeSigma
  • Overexpression of a Short Sulfonylurea Splice Variant Increases Cardiac Glucose Uptake and Uncouples Mitochondria by Regulating ROMK Activity.
Overexpression of a Short Sulfonylurea Splice Variant Increases Cardiac Glucose Uptake and Uncouples Mitochondria by Regulating ROMK Activity.
Life (Basel, Switzerland) (2023-04-28)
Sarah K El-Meanawy, Holly Dooge, Allison C Wexler, Anna C Kosmach, Lara Serban, Elizabeth A Santos, Francisco J Alvarado, Timothy A Hacker, Mohun Ramratnam
ABSTRACT
The mitochondrial splice variant of the sulfonylurea receptor (SUR2A-55) is associated with protection from myocardial ischemia-reperfusion (IR) injury, increased mitochondrial ATP sensitive K+ channel activity (mitoKATP) and altered glucose metabolism. While mitoKATP channels composed of CCDC51 and ABCB8 exist, the mitochondrial K+ pore regulated by SUR2A-55 is unknown. We explored whether SUR2A-55 regulates ROMK to form an alternate mitoKATP. We assessed glucose uptake in mice overexpressing SUR2A-55 (TGSUR2A-55) compared with WT mice during IR injury. We then examined the expression level of ROMK and the effect of ROMK modulation on mitochondrial membrane potential (Δψm) in WT and TGSUR2A-55 mice. TGSUR2A-55 had increased glucose uptake compared to WT mice during IR injury. The expression of ROMK was similar in WT compared to TGSUR2A-55 mice. ROMK inhibition hyperpolarized resting cardiomyocyte Δψm from TGSUR2A-55 mice but not from WT mice. In addition, TGSUR2A-55 and ROMK inhibitor treated WT isolated cardiomyocytes had enhanced mitochondrial uncoupling. ROMK inhibition blocked diazoxide induced Δψm depolarization and prevented preservation of Δψm from FCCP perfusion in WT and to a lesser degree TGSUR2A-55 mice. In conclusion, cardio-protection from SUR2A-55 is associated with ROMK regulation, enhanced mitochondrial uncoupling and increased glucose uptake.
Sigma-Aldrich
Anti-CCDC51 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution