MilliporeSigma
  • Effects of Mouse Kidney Parvovirus on Pharmacokinetics of Chemotherapeutics and the Adenine Model of Chronic Kidney Disease.
Effects of Mouse Kidney Parvovirus on Pharmacokinetics of Chemotherapeutics and the Adenine Model of Chronic Kidney Disease.
Comparative medicine (2023-03-28)
Amanda C Ritter, Rodolfo J Ricart Arbona, Robert S Livingston, Sébastien Monette, Neil S Lipman
ABSTRACT
Mouse kidney parvovirus (MKPV) causes inclusion body nephropathy in severely immunocompromised mice and renal interstitial inflammation in immunocompetent mice. Here we sought to determine the effects of MKPV on pre-clinical murine models that depend on renal function. To assess the effects of MKPV infection on the pharmacokinetics of 2 renally excreted chemotherapeutic agents, methotrexate and lenalidomide, we measured drug concentrations in the blood and urine of MKPV-infected or uninfected immunodeficient NOD.Cg-PrkdcscidIl2rgtm1Wjl/SzJ (NSG) and immunocompetent C57BL/6NCrl (B6) female mice. No differences in plasma pharmacokinetics were observed for lenalidomide. However, the AUC of methotrexate was 1.5-fold higher in uninfected NSG mice compared with infected NSG mice, 1.9-fold higher in infected B6 mice compared with uninfected B6 mice, and 4.3-fold higher in uninfected NSG mice compared with uninfected B6 mice. MKPV infection did not significantly affect the renal clearance of either drug. To assess effects of MKPV infection on the adenine diet model of chronic kidney disease, MKPV-infected and uninfected B6 female mice were fed a 0.2% adenine diet, and clinical and histopathologic features of disease were assessed over 8 wk. MKPV infection did not significantly alter urine chemistry results, hemogram findings, or serum concentrations of BUN, creatinine, or symmetric dimethylarginine. However, infection did influence histologic outcomes. As compared with uninfected mice, MKPV-infected mice had more interstitial lymphoplasmacytic infiltrates after 4 and 8 wk of diet consumption and less interstitial fibrosis at week 8. Macrophage infiltrates and renal tubular injury were similar between in infected and uninfected mice. These findings indicate that MKPV infection had minimal effects on the renal excretion of 2 chemotherapeutics and on serum biomarkers of renal function. However, infection significantly influenced two histologic features of the adenine diet model of chronic renal disease. MKPV-free mice are critically important in studies evaluating renal histology as an experimental outcome.
Sigma-Aldrich
Lenalidomide, ≥98% (HPLC)