MilliporeSigma
  • Development and evaluation of a novel chromium III-based compound for potential inhibition of emerging SARS-CoV-2 variants.
Development and evaluation of a novel chromium III-based compound for potential inhibition of emerging SARS-CoV-2 variants.
Heliyon (2023-10-09)
Yu-Jung Lin, Navaneethan Sundhar, Hema Sri Devi, Hsueh-Fa Pien, Shina Fong-Mei Wen, Jenn-Line Sheu, Bruce Chi-Kang Tsai, Chih-Yang Huang
ABSTRACT
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) has caused 403 million cases of coronavirus disease (COVID-19) and resulted in more than 5.7 million deaths worldwide. Extensive research has identified several potential drug treatments for COVID-19. However, the development of new compounds or therapies is necessary to prevent the emergence of drug resistance in SARS-CoV-2. In this study, a novel compound based on hexaacetotetraaquadihydroxochromium(III)diiron(III) nitrate, which contains small amounts of chromium (III), was synthesised and evaluated for its effectiveness against multiple variants of COVID-19 using both in vitro and in vivo models. This innovative compound demonstrated interference with the interaction between the spike protein of SARS-CoV-2 and angiotensin-converting enzyme 2 (ACE2). Furthermore, in vitro experiments showed that this compound downregulated the expression of ACE2 and transmembrane serine protease 2 (TMPRSS2). It also exhibited a reduction in the activity of 3-chymotrypsin-like protease (3CL) and RNA-dependent RNA polymerase (RdRp). Pretreatment with this small chromium (III)-based compound resulted in reduced ACE2-rich cell infection by various variants of SARS-CoV-2 spike protein-pseudotyped lentivirus. Finally, the compound effectively inhibited viral infection by multiple variants of SARS-CoV-2 spike protein-pseudotyped lentivirus in both the abdominal and thoracic regions of mice. In conclusion, this compound lowers the likelihood of SARS-CoV-2 entry into cells, inhibits viral maturation and replication in vitro, and reduces infection levels of multiple variants of SARS-CoV-2 spike protein-pseudotyped lentivirus in the abdomen and thorax following pretreatment. Small chromium (III)-based compounds have the potential to restrict the progression of SARS-CoV-2 infections.
Product Description
BEAS-2B Cell Line human