MilliporeSigma
  • Pre-Clinical Development of an Adenovirus Vector Based RSV and Shingles Vaccine Candidate.
Pre-Clinical Development of an Adenovirus Vector Based RSV and Shingles Vaccine Candidate.
Vaccines (2023-11-25)
Lawrence Petherbridge, Charlotte Davis, Angela Robinson, Thomas Evans, Sarah Sebastian
ABSTRACT
Respiratory syncytial virus (RSV) infection and shingles are two viral diseases that affect older adults, and a combined vaccine to protect against both could be beneficial. RSV infection causes hospitalisations and significant morbidity in both children and adults and can be fatal in the elderly. The RSV fusion (F) envelope glycoprotein induces a strong RSV-neutralising antibody response and is the target of protective immunity in the first RSV vaccine for older adults, recently approved by the FDA. An initial childhood infection with the varicella zoster virus (VZV) results in chickenpox disease, but reactivation in older adults can cause shingles. This reactivation in sensory and autonomic neurons is characterized by a skin-blistering rash that can be accompanied by prolonged pain. The approved protein-in-adjuvant shingles vaccine induces VZV glycoprotein E (gE)-fspecific antibody and CD4+ T cell responses and is highly effective. Here we report the evaluation of RSV/shingles combination vaccine candidates based on non-replicating chimpanzee adenovirus (ChAd) vectors. We confirmed the cellular and humoral immunogenicity of the vaccine vectors in mice using T cell and antibody assays. We also carried out an RSV challenge study in cotton rats which demonstrated protective efficacy following a homologous prime-boost regimen with our preferred vaccine candidate.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, F(ab′)2, HRP conjugate, Chemicon®, from goat