MilliporeSigma
  • Nanoparticles of Lactiplantibacillus plantarum K8 Reduce Staphylococcus aureus Respiratory Infection and Tumor Necrosis Factor Alpha- and Interferon Gamma-Induced Lung Inflammation.
Nanoparticles of Lactiplantibacillus plantarum K8 Reduce Staphylococcus aureus Respiratory Infection and Tumor Necrosis Factor Alpha- and Interferon Gamma-Induced Lung Inflammation.
Nutrients (2023-11-25)
Jonghyo Hong, Minseong Son, Jaeeun Sin, Hangeun Kim, Dae-Kyun Chung
ABSTRACT
Multiple studies have confirmed that Lactiplantibacillus plantarum has beneficial effects in respiratory diseases, including respiratory tract infections, asthma, and chronic obstructive pulmonary disease. However, the role of L. plantarum lysates in respiratory diseases is unclear. Staphylococcus aureus infects the lungs of mice, recruits immune cells, and induces structural changes in alveoli. Lung diseases can be further aggravated by inflammatory cytokines such as CCL2 and interleukin (IL)-6. In in vivo studies, L. plantarum K8 nanoparticles (K8NPs) restored lung function and prevented lung damage caused by S. aureus infection. They inhibited the S. aureus infection and the infiltration of immune cells and prevented the increase in goblet cell numbers in the lungs of S. aureus-infected mice. K8NPs suppressed the expression of CCL2 and IL-6, which were increased by the combination treatment of tumor necrosis factor alpha and interferon gamma (TI), in a dose-dependent manner. In in vitro studies, the anti-inflammatory effect of K8NPs in TI-treated A549 cells and TI-injected mice occurred through the reduction in activated mitogen-activated protein kinases and nuclear factor kappa-B. These findings suggest that the efficacy of K8NPs in controlling respiratory inflammation and infection can be used to develop functional materials that can prevent or alleviate respiratory diseases.
Sigma-Aldrich
Anti-F4/80 antibody, Rabbit monoclonal, recombinant, expressed in proprietary host, clone SP115, affinity isolated antibody