MilliporeSigma
  • MYOD-SKP2 axis boosts tumorigenesis in fusion negative rhabdomyosarcoma by preventing differentiation through p57Kip2 targeting.
MYOD-SKP2 axis boosts tumorigenesis in fusion negative rhabdomyosarcoma by preventing differentiation through p57Kip2 targeting.
Nature communications (2023-12-16)
Silvia Pomella, Matteo Cassandri, Lucrezia D'Archivio, Antonella Porrazzo, Cristina Cossetti, Doris Phelps, Clara Perrone, Michele Pezzella, Antonella Cardinale, Marco Wachtel, Sara Aloisi, David Milewski, Marta Colletti, Prethish Sreenivas, Zoë S Walters, Giovanni Barillari, Angela Di Giannatale, Giuseppe Maria Milano, Cristiano De Stefanis, Rita Alaggio, Sonia Rodriguez-Rodriguez, Nadia Carlesso, Christopher R Vakoc, Enrico Velardi, Beat W Schafer, Ernesto Guccione, Susanne A Gatz, Ajla Wasti, Marielle Yohe, Myron Ignatius, Concetta Quintarelli, Janet Shipley, Lucio Miele, Javed Khan, Peter J Houghton, Francesco Marampon, Berkley E Gryder, Biagio De Angelis, Franco Locatelli, Rossella Rota
ABSTRACT
Rhabdomyosarcomas (RMS) are pediatric mesenchymal-derived malignancies encompassing PAX3/7-FOXO1 Fusion Positive (FP)-RMS, and Fusion Negative (FN)-RMS with frequent RAS pathway mutations. RMS express the master myogenic transcription factor MYOD that, whilst essential for survival, cannot support differentiation. Here we discover SKP2, an oncogenic E3-ubiquitin ligase, as a critical pro-tumorigenic driver in FN-RMS. We show that SKP2 is overexpressed in RMS through the binding of MYOD to an intronic enhancer. SKP2 in FN-RMS promotes cell cycle progression and prevents differentiation by directly targeting p27Kip1 and p57Kip2, respectively. SKP2 depletion unlocks a partly MYOD-dependent myogenic transcriptional program and strongly affects stemness and tumorigenic features and prevents in vivo tumor growth. These effects are mirrored by the investigational NEDDylation inhibitor MLN4924. Results demonstrate a crucial crosstalk between transcriptional and post-translational mechanisms through the MYOD-SKP2 axis that contributes to tumorigenesis in FN-RMS. Finally, NEDDylation inhibition is identified as a potential therapeutic vulnerability in FN-RMS.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys27) Antibody, clone RM172, clone RM172, from rabbit