MilliporeSigma
  • Enhanced production of mesencephalic dopaminergic neurons from lineage-restricted human undifferentiated stem cells.
Enhanced production of mesencephalic dopaminergic neurons from lineage-restricted human undifferentiated stem cells.
Nature communications (2023-12-06)
Muyesier Maimaitili, Muwan Chen, Fabia Febbraro, Ekin Ucuncu, Rachel Kelly, Jonathan Christos Niclis, Josefine Rågård Christiansen, Noëmie Mermet-Joret, Dragos Niculescu, Johanne Lauritsen, Angelo Iannielli, Ida H Klæstrup, Uffe Birk Jensen, Per Qvist, Sadegh Nabavi, Vania Broccoli, Anders Nykjær, Marina Romero-Ramos, Mark Denham
ABSTRACT
Current differentiation protocols for generating mesencephalic dopaminergic (mesDA) neurons from human pluripotent stem cells result in grafts containing only a small proportion of mesDA neurons when transplanted in vivo. In this study, we develop lineage-restricted undifferentiated stem cells (LR-USCs) from pluripotent stem cells, which enhances their potential for differentiating into caudal midbrain floor plate progenitors and mesDA neurons. Using a ventral midbrain protocol, 69% of LR-USCs become bona fide caudal midbrain floor plate progenitors, compared to only 25% of human embryonic stem cells (hESCs). Importantly, LR-USCs generate significantly more mesDA neurons under midbrain and hindbrain conditions in vitro and in vivo. We demonstrate that midbrain-patterned LR-USC progenitors transplanted into 6-hydroxydopamine-lesioned rats restore function in a clinically relevant non-pharmacological behavioral test, whereas midbrain-patterned hESC-derived progenitors do not. This strategy demonstrates how lineage restriction can prevent the development of undesirable lineages and enhance the conditions necessary for mesDA neuron generation.
Sigma-Aldrich
Anti-GABA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-LMX-1 Antibody, serum, from rabbit
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®
Sigma-Aldrich
Anti-Serotonin antibody produced in rabbit, whole antiserum