MilliporeSigma
  • Injury-specific factors in the cerebrospinal fluid regulate astrocyte plasticity in the human brain.
Injury-specific factors in the cerebrospinal fluid regulate astrocyte plasticity in the human brain.
Nature medicine (2023-12-09)
Swetlana Sirko, Christian Schichor, Patrizia Della Vecchia, Fabian Metzger, Giovanna Sonsalla, Tatiana Simon, Martina Bürkle, Sofia Kalpazidou, Jovica Ninkovic, Giacomo Masserdotti, Jean-Frederic Sauniere, Valentina Iacobelli, Stefano Iacobelli, Claire Delbridge, Stefanie M Hauck, Jörg-Christian Tonn, Magdalena Götz
ABSTRACT
The glial environment influences neurological disease progression, yet much of our knowledge still relies on preclinical animal studies, especially regarding astrocyte heterogeneity. In murine models of traumatic brain injury, beneficial functions of proliferating reactive astrocytes on disease outcome have been unraveled, but little is known regarding if and when they are present in human brain pathology. Here we examined a broad spectrum of pathologies with and without intracerebral hemorrhage and found a striking correlation between lesions involving blood-brain barrier rupture and astrocyte proliferation that was further corroborated in an assay probing for neural stem cell potential. Most importantly, proteomic analysis unraveled a crucial signaling pathway regulating this astrocyte plasticity with GALECTIN3 as a novel marker for proliferating astrocytes and the GALECTIN3-binding protein LGALS3BP as a functional hub mediating astrocyte proliferation and neurosphere formation. Taken together, this work identifies a therapeutically relevant astrocyte response and their molecular regulators in different pathologies affecting the human cerebral cortex.
Sigma-Aldrich
Monoclonal Anti-S-100 (β-Subunit) antibody produced in mouse, clone SH-B1, ascites fluid
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Oligodendrocyte Marker O4 antibody produced in mouse, clone O4, purified immunoglobulin, lyophilized powder
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
DAPI, for nucleic acid staining