MilliporeSigma
  • Combinations of EGFR and MET inhibitors reduce proliferation and invasiveness of mucosal melanoma cells.
Combinations of EGFR and MET inhibitors reduce proliferation and invasiveness of mucosal melanoma cells.
Journal of cellular and molecular medicine (2023-09-08)
Aleksandra Simiczyjew, Justyna Wądzyńska, Magdalena Kot, Marcin Ziętek, Rafał Matkowski, Mai P Hoang, Piotr Donizy, Dorota Nowak
ABSTRACT
Mucosal melanoma (MM) is a very rare and aggressive type of cancer for which immunotherapy or targeted therapy such as BRAF/MEK inhibitors, used in cutaneous melanoma, usually fail. Due to our earlier experience showing the high effectiveness of epidermal growth factor receptor (EGFR) and hepatocyte growth factor receptor (MET) inhibitors in reducing the activation of the MAPK and PI3K/AKT signalling pathways, we aim to test whether these drugs would also be effective for mucosal melanoma. Cells representing two commercially available mucosal melanoma cell lines (GAK and HMVII) and one cell line obtained from a patient's vaginal melanoma were treated with MET or EGFR inhibitors, or combinations of these agents. The dual-inhibitor treatment strategy resulted in a decrease of cell proliferation, migration and invasion. Moreover, combinations of inhibitors led to reduction of pEGFR/EGFR and pMET/MET ratio and downregulation of PI3K/AKT and MEK/ERK1/2-based signalling pathways. Our findings indicate a potential therapeutic strategy based on EGFR and MET inhibitors in mucosal melanoma, which should be further evaluated in vivo and in clinical experiments. They also suggest that targeting multiple receptor tyrosine kinases may block signalling crosstalk and possibly delay the appearance of resistance to kinase inhibitors in mucosal melanoma cells.
Sigma-Aldrich
Anti-Melan-A antibody, Mouse monoclonal, clone A103, purified from hybridoma cell culture
Sigma-Aldrich
Anti-phospho-c-Met (pTyr1230/pTyr1234/pTyr1235) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MAGEC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution