MilliporeSigma
  • LEF1 Drives a Central Memory Program and Supports Antitumor Activity of Natural Killer T Cells.
LEF1 Drives a Central Memory Program and Supports Antitumor Activity of Natural Killer T Cells.
Cancer immunology research (2022-12-10)
Ho Ngai, Gabriel A Barragan, Gengwen Tian, Julien C Balzeau, Chunchao Zhang, Amy N Courtney, Linjie Guo, Xin Xu, Michael S Wood, Janice M Drabek, Thorsten Demberg, Caroline M Sands, Cynthia N Chauvin-Fleurence, Erica J Di Pierro, Jeffrey M Rosen, Leonid S Metelitsa
ABSTRACT
Vα24-invariant natural killer T cells (NKT) possess innate antitumor properties that can be exploited for cancer immunotherapy. We have shown previously that the CD62L+ central memory-like subset of these cells drives the in vivo antitumor activity of NKTs, but molecular mediators of NKT central memory differentiation remain unknown. Here, we demonstrate that relative to CD62L- cells, CD62L+ NKTs express a higher level of the gene encoding the Wnt/β-catenin transcription factor lymphoid enhancer binding factor 1 (LEF1) and maintain active Wnt/β-catenin signaling. CRISPR/Cas9-mediated LEF1 knockout reduced CD62L+ frequency after antigenic stimulation, whereas Wnt/β-catenin activator Wnt3a ligand increased CD62L+ frequency. LEF1 overexpression promoted NKT expansion and limited exhaustion following serial tumor challenge and was sufficient to induce a central memory-like transcriptional program in NKTs. In mice, NKTs expressing a GD2-specific chimeric-antigen receptor (CAR) with LEF1 demonstrated superior control of neuroblastoma xenograft tumors compared with control CAR-NKTs. These results identify LEF1 as a transcriptional activator of the NKT central memory program and advance development of NKT cell-based immunotherapy. See related Spotlight by Van Kaer, p. 144.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
GSK-3β Inhibitor XII, TWS119, The GSK-3β Inhibitor XII, TWS119, also referenced under CAS 601514-19-6, controls the biological activity of GSK-3β. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.