MilliporeSigma
  • Evolution of the electrochemical interface in sodium ion batteries with ether electrolytes.
Evolution of the electrochemical interface in sodium ion batteries with ether electrolytes.
Nature communications (2019-02-15)
Kaikai Li, Jun Zhang, Dongmei Lin, Da-Wei Wang, Baohua Li, Wei Lv, Sheng Sun, Yan-Bing He, Feiyu Kang, Quan-Hong Yang, Limin Zhou, Tong-Yi Zhang
ABSTRACT
Ether based electrolytes have surfaced as alternatives to conventional carbonates allowing for enhanced electrochemical performance of sodium-ion batteries; however, the primary source of the improvement remains poorly understood. Here we show that coupling titanium dioxide and other anode materials with diglyme does enable higher efficiency and reversible capacity than those for the combination involving ester electrolytes. Importantly, the electrolyte dependent performance is revealed to be the result of the different structural evolution induced by a varied sodiation depth. A suit of characterizations show that the energy barrier to charge transfer at the interface between electrolyte and electrode is the factor that dominates the interfacial electrochemical characteristics and therefore the energy storage properties. Our study proposes a reliable parameter to assess the intricate sodiation dynamics in sodium-ion batteries and could guide the design of aprotic electrolytes for next generation rechargeable batteries.
Sigma-Aldrich
Sodium triflate solution, in Bis(2-methoxyethyl) ether, 1.0 M NaTfO in diglyme, battery grade