MilliporeSigma
  • LAMP-2A ablation in hippocampal CA1 astrocytes confers cerebroprotection and ameliorates neuronal injury after global brain ischemia.
LAMP-2A ablation in hippocampal CA1 astrocytes confers cerebroprotection and ameliorates neuronal injury after global brain ischemia.
Brain pathology (Zurich, Switzerland) (2022-09-06)
Han-Yu Fu, Yang Cui, Qiao Li, Ding Wang, Hui Li, Long Yang, De-Juan Wang, Jing-Wei Zhou
ABSTRACT
Reactive astrogliosis and neuronal death are major features of brain tissue damage after transient global cerebral ischemia/reperfusion (I/R). The CA1 subfield in the hippocampus is particularly susceptible to cell death after I/R. Recently, attention has focused on the relationship between the autophagy-lysosomal pathway and cerebral ischemia. Lysosomal-associated membrane protein type-2A (LAMP-2A) is a key protein in chaperone-mediated autophagy (CMA). However, LAMP-2A expression in astrocytes of the hippocampus and its influence on brain injury following I/R remain unknown. Here, we show that LAMP-2A is elevated in astrocytes of the CA1 hippocampal subfield after I/R and in primary cultured astrocytes after transient oxygen-glucose deprivation (OGD). Conditional LAMP-2A knockdown in CA1 astrocytes inhibited astrocyte activation and prevented neuronal death by inhibiting the mitochondrial pathway of apoptosis after I/R, suggesting that elevated astrocytic LAMP-2A contributes to regional ischemic vulnerability. Furthermore, astrocytic LAMP-2A ablation ameliorated the spatial learning and memory deficits caused by I/R. Conditional astrocytic LAMP-2A knockdown also prevented the loss of hippocampal synapses and dendritic spines, improved the synaptic ultrastructure, and inhibited the reduced expression of synaptic proteins after ischemia. Thus, our findings demonstrate that astrocytic LAMP-2A expression increases upon I/R and that LAMP-2A ablation specifically in hippocampal astrocytes contributes to cerebroprotection, suggesting a novel neuroprotective strategy for patients with global ischemia.
Sigma-Aldrich
Anti-LAMP2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-PSD-95 (Ab-2) Mouse mAb (7E3-1B8), liquid, clone 7E3-1B8, Calbiochem®