MilliporeSigma
  • Tau-induced deficits in nonsense-mediated mRNA decay contribute to neurodegeneration.
Tau-induced deficits in nonsense-mediated mRNA decay contribute to neurodegeneration.
Alzheimer's & dementia : the journal of the Alzheimer's Association (2022-04-14)
Gabrielle Zuniga, Simon Levy, Paulino Ramirez, Jasmine De Mange, Elias Gonzalez, Maria Gamez, Bess Frost
ABSTRACT
While brains of patients with Alzheimer's disease and related tauopathies have evidence of altered RNA processing, we lack a mechanistic understanding of how altered RNA processing arises in these disorders and if such changes are causally linked to neurodegeneration. Using Drosophila melanogaster models of tauopathy, we find that overall activity of nonsense-mediated mRNA decay (NMD), a key RNA quality-control mechanism, is reduced. Genetic manipulation of NMD machinery significantly modifies tau-induced neurotoxicity, suggesting that deficits in NMD are causally linked to neurodegeneration. Mechanistically, we find that deficits in NMD are a consequence of aberrant RNA export and RNA accumulation within nuclear envelope invaginations in tauopathy. We identify a pharmacological activator of NMD that suppresses neurodegeneration in tau transgenic Drosophila, indicating that tau-induced deficits in RNA quality control are druggable. Our studies suggest that NMD activators should be explored for their potential therapeutic value to patients with tauopathies.
Sigma-Aldrich
Tranilast, ≥98% (HPLC), powder