MilliporeSigma
  • Water extract of frankincense and myrrh inhibits liver cancer progression and epithelial‑mesenchymal transition through Wnt/β‑catenin signaling.
Water extract of frankincense and myrrh inhibits liver cancer progression and epithelial‑mesenchymal transition through Wnt/β‑catenin signaling.
Molecular and clinical oncology (2023-09-18)
Xian Lu, Jialei Mao, Yaodong Wang, Yonggang Huang, Maolin Gu
ABSTRACT
Wnt/β-catenin signaling is associated with epithelial-mesenchymal transformation (EMT), which serves an important role in hepatocellular carcinoma (HCC) invasion and metastasis. Frankincense and myrrh (FM) are antitumor agents commonly used in clinical practice. The present study aimed to investigate the effect and mechanism of water extract of FM on the progression of liver cancer cells. FM was applied to study its effects on HCC cell proliferation. Cell migration and invasion were evaluated by wound healing and Transwell assays. In addition, western blot was used to study the protein levels associated with EMT and Wnt/β-catenin signaling. The nuclear translocation of β-catenin was detected by immunofluorescence assay. A non-toxic dose of FM significantly inhibited invasion and metastasis of liver cancer cells. Furthermore, FM promoted expression of EMT marker E-cadherin, while decreasing expression of vimentin and N-cadherin. Finally, the protein and the nuclear staining levels of Disheveled 2 and β-catenin were both suppressed by water extract of FM. The water extract of FM inhibited the migration and invasion of liver cancer cells and inhibited EMT by suppressing activation of the Wnt/β-catenin signaling pathway.
Sigma-Aldrich
Wnt Agonist II, SKL2001, The Wnt Agonist II, SKL2001 controls the biological activity of Wnt. This small molecule/inhibitor is primarily used for Biochemicals applications.