MilliporeSigma
  • Mitochondrial control of microglial phagocytosis by the translocator protein and hexokinase 2 in Alzheimer's disease.
Mitochondrial control of microglial phagocytosis by the translocator protein and hexokinase 2 in Alzheimer's disease.
Proceedings of the National Academy of Sciences of the United States of America (2023-02-15)
Lauren H Fairley, Kei Onn Lai, Jia Hui Wong, Wei Jing Chong, Anselm Salvatore Vincent, Giuseppe D'Agostino, Xiaoting Wu, Roshan R Naik, Anusha Jayaraman, Sarah R Langley, Christiane Ruedl, Anna M Barron
ABSTRACT
Microglial phagocytosis is an energetically demanding process that plays a critical role in the removal of toxic protein aggregates in Alzheimer's disease (AD). Recent evidence indicates that a switch in energy production from mitochondrial respiration to glycolysis disrupts this important protective microglial function and may provide therapeutic targets for AD. Here, we demonstrate that the translocator protein (TSPO) and a member of its mitochondrial complex, hexokinase-2 (HK), play critical roles in microglial respiratory-glycolytic metabolism and phagocytosis. Pharmacological and genetic loss-of-function experiments showed that TSPO is critical for microglial respiratory metabolism and energy supply for phagocytosis, and its expression is enriched in phagocytic microglia of AD mice. Meanwhile, HK controlled glycolytic metabolism and phagocytosis via mitochondrial binding or displacement. In cultured microglia, TSPO deletion impaired mitochondrial respiration and increased mitochondrial recruitment of HK, inducing a switch to glycolysis and reducing phagocytosis. To determine the functional significance of mitochondrial HK recruitment, we developed an optogenetic tool for reversible control of HK localization. Displacement of mitochondrial HK inhibited glycolysis and improved phagocytosis in TSPO-knockout microglia. Mitochondrial HK recruitment also coordinated the inflammatory switch to glycolysis that occurs in response to lipopolysaccharide in normal microglia. Interestingly, cytosolic HK increased phagocytosis independent of its metabolic activity, indicating an immune signaling function. Alzheimer's beta amyloid drastically stimulated mitochondrial HK recruitment in cultured microglia, which may contribute to microglial dysfunction in AD. Thus, targeting mitochondrial HK may offer an immunotherapeutic approach to promote phagocytic microglial function in AD.
Sigma-Aldrich
Anti-Amyloid βA4, clone 1E8 (Amino Terminus) Antibody, clone 1E8, 1 mg/mL, from mouse