MilliporeSigma
  • Lymphatics and fibroblasts support intestinal stem cells in homeostasis and injury.
Lymphatics and fibroblasts support intestinal stem cells in homeostasis and injury.
Cell stem cell (2022-08-06)
Norihiro Goto, Saori Goto, Shinya Imada, Sahar Hosseini, Vikram Deshpande, Ömer H Yilmaz
ABSTRACT
Lgr5+ intestinal stem cells (ISCs) depend on niche factors for their proper function. However, the source of these ISC niche factors and how they support ISCs in vivo remain controversial. Here, we report that ISCs depend on lymphatic endothelial cells (LECs) and RSPO3+GREM1+ fibroblasts (RGFs). In the intestine and colon, LECs are surrounded by RGFs and are located near ISCs at the crypt base. Both LECs and RGFs provide the critical ISC niche factor RSPO3 to support ISCs, where RSPO3 loss in both cell types drastically compromises ISC numbers, villi length, and repair after injury. In response to injury, LEC and RGF numbers expand and produce greater amounts of RSPO3 and other growth/angiocrine factors to foster intestinal repair. We propose that LECs represent a novel niche component for ISCs, which together with RGFs serve as the major in vivo RSPO3 source for ISCs in homeostasis and injury-mediated regeneration.
Sigma-Aldrich
Anti-LYVE1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)