MilliporeSigma
  • Protein nanopore reveals the renin-angiotensin system crosstalk with single-amino-acid resolution.
Protein nanopore reveals the renin-angiotensin system crosstalk with single-amino-acid resolution.
Nature chemistry (2023-02-23)
Jie Jiang, Meng-Yin Li, Xue-Yuan Wu, Yi-Lun Ying, Huan-Xing Han, Yi-Tao Long
ABSTRACT
The discovery of crosstalk effects on the renin-angiotensin system (RAS) is limited by the lack of approaches to quantitatively monitor, in real time, multiple components with subtle differences and short half-lives. Here we report a nanopore framework to quantitatively determine the effect of the hidden crosstalk between angiotensin-converting enzyme (ACE) and angiotensin-converting enzyme 2 (ACE2) on RAS. By developing an engineered aerolysin nanopore capable of single-amino-acid resolution, we show that the ACE can be selectively inhibited by ACE2 to prevent cleavage of angiotensin I, even when the concentration of ACE is more than 30-fold higher than that of ACE2. We also show that the activity of ACE2 for cleaving angiotensin peptides is clearly suppressed by the spike protein of SARS-CoV-2. This leads to the relaxation of ACE and the increased probability of accumulation of the principal effector angiotensin II. The spike protein of the SARS-CoV-2 Delta variant is demonstrated to have a much greater impact on the crosstalk than the wild type.
Sigma-Aldrich
Trypsin Agarose, buffered aqueous suspension, from bovine pancreas trypsin