MilliporeSigma
  • Innate Immune Status of Glia Modulates Prion Propagation in Early Stage of Infection.
Innate Immune Status of Glia Modulates Prion Propagation in Early Stage of Infection.
Cells (2023-07-29)
Sang-Gyun Kang, Chiye Kim, Judd Aiken, Debbie McKenzie
ABSTRACT
Prion diseases are progressive neurodegenerative disorders affecting humans and various mammals. The prominent neuropathological change in prion-affected brains is neuroinflammation, histopathologically characterized by reactive gliosis surrounding prion deposition. The cause and effect of these cellular responses are still unclear. Here we investigate the impact of innate immune responses on prion replication using in vitro cell culture models. Hamster-adapted transmissible mink encephalopathy prions, hyper (HY) and drowsy (DY) strains, were assayed for accumulation of pathogenic prion protein (PrPSc) in primary glial cultures derived from 8-day-old hamster pups. The kinetics of PrPSc accumulation largely depended on prion strain and brain regions from where glial cells originated. Glial cells derived from the cerebellum were susceptible to HY, but resistant to DY strain as determined by western blot analysis, immunocytochemistry, and animal bioassay. Glial cells from the cerebral cortex were, however, refractory to both strains. PrPSc accumulation was affected by innate immune modulators. Priming glial cells with lipopolysaccharide decreased prion replication, whereas pre-treatment with dexamethasone, inhibiting innate immunity, increased susceptibility to DY infection. Our results suggest that neuroinflammation resulting from prion infection is a response to resolve and/or prevent prion propagation in the brain. It implies a therapeutic potential of innate immune modulation in the early stages of prion disease.
Sigma-Aldrich
Anti-Potassium Channel KCa2.3, (C-term) (Small-conductance Ca2+-activated Potassium Channel 3) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-Glutamine Synthetase Antibody, clone GS-6, clone GS-6, Chemicon®, from mouse
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse