MilliporeSigma
Malonyl-CoA is a conserved endogenous ATP-competitive mTORC1 inhibitor.
Nature cell biology (2023-08-11)
Raffaele Nicastro, Laura Brohée, Josephine Alba, Julian Nüchel, Gianluca Figlia, Stefanie Kipschull, Peter Gollwitzer, Jesus Romero-Pozuelo, Stephanie A Fernandes, Andreas Lamprakis, Stefano Vanni, Aurelio A Teleman, Claudio De Virgilio, Constantinos Demetriades
ABSTRACT
Cell growth is regulated by the mammalian/mechanistic target of rapamycin complex 1 (mTORC1), which functions both as a nutrient sensor and a master controller of virtually all biosynthetic pathways. This ensures that cells are metabolically active only when conditions are optimal for growth. Notably, although mTORC1 is known to regulate fatty acid biosynthesis, how and whether the cellular lipid biosynthetic capacity signals back to fine-tune mTORC1 activity remains poorly understood. Here we show that mTORC1 senses the capacity of a cell to synthesise fatty acids by detecting the levels of malonyl-CoA, an intermediate of this biosynthetic pathway. We find that, in both yeast and mammalian cells, this regulation is direct, with malonyl-CoA binding to the mTOR catalytic pocket and acting as a specific ATP-competitive inhibitor. When fatty acid synthase (FASN) is downregulated/inhibited, elevated malonyl-CoA levels are channelled to proximal mTOR molecules that form direct protein-protein interactions with acetyl-CoA carboxylase 1 (ACC1) and FASN. Our findings represent a conserved and unique homeostatic mechanism whereby impaired fatty acid biogenesis leads to reduced mTORC1 activity to coordinately link this metabolic pathway to the overall cellular biosynthetic output. Moreover, they reveal the existence of a physiological metabolite that directly inhibits the activity of a signalling kinase in mammalian cells by competing with ATP for binding.
Sigma-Aldrich
Fasnall benzenesulfonate salt, ≥98% (HPLC)
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Cerulenin, ≥98% (HPLC), from Cephalosporium caerulens