MilliporeSigma
  • The pathogenesis of cryptoglandular anal fistula: New insight into the immunological profile.
The pathogenesis of cryptoglandular anal fistula: New insight into the immunological profile.
Colorectal disease : the official journal of the Association of Coloproctology of Great Britain and Ireland (2022-08-03)
Francesco Litta, Andrea Papait, Donatella Lucchetti, Serafina Farigu, Angelo Parello, Claudio Ricciardi Tenore, Paola Campennì, Antonietta Rosa Silini, Maria Cristina Giustiniani, Ornella Parolini, Alessandro Sgambato, Carlo Ratto
ABSTRACT
The aetiology of cryptoglandular anal fistula (AF) is poorly understood. Evidence suggests that persistence and/or recurrence of the disease is more related to inflammatory than infectious factors. The aim of this study was to investigate the immune profile of cryptoglandular AF and to perform a histopathological characterization. Fistulectomy was performed in all patients; healthy ischioanal fat from the same patients was used as a control. Samples were evaluated by the Luminex xMAP system for the detection of 27 analytes. AF tissues were analysed using immunofluorescence. Staining was performed using primary antibodies to identify M1 inflammatory and M2 anti-inflammatory macrophages. Selective staining of total T lymphocytes and different T lymphocyte subsets was performed. Twenty patients with AF underwent a fistulectomy. Specific cytokine pathways differentiated AF from healthy tissue: pro-inflammatory cytokines interleukin (IL)-1β, IL-4, IL-8 and IL-17 and the anti-inflammatory cytokine IL-10 were overexpressed in AF compared with controls. Chemokines involved in macrophage recruitment (CCL2, CCL3, CCL4) were higher in AF than in healthy fatty tissue. Moreover, we showed that Tc17 cells characterize AF patients, thus confirming the enzyme-linked immunosorbent assay data. Furthermore, elevated infiltration of CD68+ myeloid cells and a reduction of the M1/M2 ratio characterize AF patients. A combination of inflammatory cytokines, chemokines and growth factors reside in the wound microenvironment of AF patients. For the first time an important prevalence of Tc17 cells and a reduction in the M1/M2 ratio was observed, thus suggesting new insights into the immunological characterization of AF patients.
Sigma-Aldrich
Anti-iNOS/NOS II Antibody, NT, from rabbit, purified by affinity chromatography