MilliporeSigma
  • Molecular basis of GDF15 induction and suppression by drugs in cardiomyocytes and cancer cells toward precision medicine.
Molecular basis of GDF15 induction and suppression by drugs in cardiomyocytes and cancer cells toward precision medicine.
Scientific reports (2023-07-27)
Lisa-Maria Winter, Diana Reinhardt, Ariane Schatter, Vivien Tissen, Heike Wiora, Daniel Gerlach, Ulrike Tontsch-Grunt, Florian Colbatzky, Birgit Stierstorfer, Seong-Wook Yun
ABSTRACT
GDF15 has recently emerged as a key driver of the development of various disease conditions including cancer cachexia. Not only the tumor itself but also adverse effects of chemotherapy have been reported to contribute to increased GDF15. Although regulation of GDF15 transcription by BET domain has recently been reported, the molecular mechanisms of GDF15 gene regulation by drugs are still unknown, leaving uncertainty about the safe and effective therapeutic strategies targeting GDF15. We screened various cardiotoxic drugs and BET inhibitors for their effects on GDF15 regulation in human cardiomyocytes and cancer cell lines and analyzed in-house and public gene signature databases. We found that DNA damaging drugs induce GDF15 in cardiomyocytes more strongly than drugs with other modes of action. In cancer cells, GDF15 induction varied depending on drug- and cell type-specific gene signatures including mutations in PI3KCA, TP53, BRAF and MUC16. GDF15 suppression by BET inhibition is particularly effective in cancer cells with low activity of the PI3K/Akt axis and high extracellular concentrations of pantothenate. Our findings provide insights that the risk for GDF15 overexpression and concomitant cachexia can be reduced by a personalized selection of anticancer drugs and patients for precision medicine.
Sigma-Aldrich
GSK1210151A, ≥98% (HPLC)
Sigma-Aldrich
CPI203, ≥98% (HPLC)