MilliporeSigma
  • Biochemical and Functional Characterization of the Three Zebrafish Transglutaminases 2.
Biochemical and Functional Characterization of the Three Zebrafish Transglutaminases 2.
International journal of molecular sciences (2023-08-12)
Manuel Lisetto, Mariagiulia Fattorini, Andrea Lanza, Marco Gerdol, Martin Griffin, Zhuo Wang, Fortunato Ferrara, Daniele Sblattero
ABSTRACT
Transglutaminase 2 (TG2) is a multifunctional protein widely distributed in various tissues and involved in many physiological and pathological processes. However, its actual role in biological processes is often controversial as TG2 shows different effects in these processes depending on its localization, cell type, or experimental conditions. We characterized the enzymatic and functional properties of TG2 proteins expressed in Danio rerio (zebrafish) to provide the basis for using this established animal model as a reliable tool to characterize TG2 functions in vivo. We confirmed the existence of three genes orthologous to human TG2 (zTGs2) in the zebrafish genome and their expression and function during embryonic development. We produced and purified the zTGs2s as recombinant proteins and showed that, like the human enzyme, zTGs2 catalyzes a Ca2+ dependent transamidation reaction that can be inhibited with TG2-specific inhibitors. In a cell model of human fibroblasts, we also demonstrated that zTGs2 can mediate RGD-independent cell adhesion in the extracellular environment. Finally, we transfected and selected zTGs2-overexpressing HEK293 cells and demonstrated that intracellular zTGs2 plays a very comparable protective/damaging role in the apoptotic process, as hTG2. Overall, our results suggest that zTGs2 proteins behave very similarly to the human ortholog and pave the way for future in vivo studies of TG2 functions in zebrafish.
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Transglutaminase 2 Inhibitor, ZDON, A cell-permeable, peptide-based, irreversible inhibitor of transglutaminase 2 (TG2; IC₅₀ = 150 nM for recombinant TG2). Reacts with the active site cysteine of TG2.
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks