MilliporeSigma
  • Involvement of CaV 2.2 channels and α2 δ-1 in homeostatic synaptic plasticity in cultured hippocampal neurons.
Involvement of CaV 2.2 channels and α2 δ-1 in homeostatic synaptic plasticity in cultured hippocampal neurons.
The Journal of physiology (2022-11-16)
Kjara S Pilch, Krishma H Ramgoolam, Annette C Dolphin
ABSTRACT
In the mammalian brain, presynaptic CaV 2 channels play a pivotal role in synaptic transmission by mediating fast neurotransmitter exocytosis via influx of Ca2+ into the active zone of presynaptic terminals. However, the distribution and modulation of CaV 2.2 channels at plastic hippocampal synapses remains to be elucidated. Here, we assess CaV 2.2 channels during homeostatic synaptic plasticity, a compensatory form of homeostatic control preventing excessive or insufficient neuronal activity during which extensive active zone remodelling has been described. We show that chronic silencing of neuronal activity in mature hippocampal cultures resulted in elevated presynaptic Ca2+ transients, mediated by increased levels of CaV 2.2 channels at the presynaptic site. This work focused further on the role of α2 δ-1 subunits, important regulators of synaptic transmission and CaV 2.2 channel abundance at the presynaptic membrane. We found that α2 δ-1 overexpression reduces the contribution of CaV 2.2 channels to total Ca2+ flux without altering the amplitude of the Ca2+ transients. Levels of endogenous α2 δ-1 decreased during homeostatic synaptic plasticity, whereas the overexpression of α2 δ-1 prevented homeostatic synaptic plasticity in hippocampal neurons. Together, this study reveals a key role for CaV 2.2 channels and novel roles for α2 δ-1 during synaptic plastic adaptation. KEY POINTS: The roles of CaV 2.2 channels and α2 δ-1 in homeostatic synaptic plasticity in hippocampal neurons in culture were examined. Chronic silencing of neuronal activity resulted in elevated presynaptic Ca2+ transients, mediated by increased levels of CaV 2.2 channels at presynaptic sites. The level of endogenous α2 δ-1 decreased during homeostatic synaptic plasticity, whereas overexpression of α2 δ-1 prevented homeostatic synaptic plasticity in hippocampal neurons. Together, this study reveals a key role for CaV 2.2 channels and novel roles for α2 δ-1 during synaptic plastic adaptation.
Sigma-Aldrich
Anti-Calcium Channel (α2/δ-1 Subunit) antibody produced in rabbit, affinity isolated antibody, lyophilized powder