MilliporeSigma
Improving Gene Therapy for Angelman Syndrome with Secreted Human UBE3A.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics (2022-05-10)
Austin W Nenninger, Matthew Willman, Jonathan Willman, Emma Stewart, Philippe Mesidor, Michelle Novoa, Nicole K Morrill, Luis Alvarez, Aurélie Joly-Amado, Melinda M Peters, Danielle Gulick, Kevin R Nash
ABSTRACT
The rare genetic neurodevelopmental disease Angelman syndrome (AS) is caused by the loss of function of UBE3A, a ubiquitin ligase. The disease results in a lifetime of severe symptoms, including intellectual disability and motor impairments for which there are no effective treatments. One avenue of treatment for AS is the use of gene therapy to reintroduce a functional copy of the UBE3A gene. Our group had previously shown that recombinant adeno-associated virus (rAAV) expressing mouse Ube3a could rescue deficits in a mouse model of AS. Here, we expand on this work and show that this approach could be successfully replicated in a second AS model using the human UBE3A gene. Furthermore, we address the challenge of limited vector distribution in the brain by developing a novel modified form of UBE3A. This modified protein, termed STUB, was designed with a secretion signal and a cell-penetrating peptide. This allowed transduced cells to act as factories for the production of UBE3A protein that could be taken up by neighboring non-transduced cells, thus increasing the number of neurons receiving the therapeutic protein. Combining this construct with intracerebroventricular injections to maximize rAAV distribution within the brain, we demonstrate that this novel approach improves the recovery of behavioral and electrophysiological deficits in the AS rat model. More importantly, a comparison of rAAV-STUB to a rAAV expressing the normal human UBE3A gene showed that STUB was a more effective therapeutic. These data suggest that rAAV-STUB is a new potential approach for the treatment of AS.
Sigma-Aldrich
Monoclonal Anti-UBE3A antibody produced in mouse, clone 3E5, purified immunoglobulin, buffered aqueous solution