MilliporeSigma
  • Telomerase inhibitor MST-312 and quercetin synergistically inhibit cancer cell proliferation by promoting DNA damage.
Telomerase inhibitor MST-312 and quercetin synergistically inhibit cancer cell proliferation by promoting DNA damage.
Translational oncology (2022-10-25)
Stina George Fernandes, Kavita Gala, Ekta Khattar
ABSTRACT
Quercetin is a natural flavonoid with well-established anti-proliferative activities against a variety of cancers. Telomerase inhibitor MST-312 also exhibits anti-proliferative effect on various cancer cells independent of its effect on telomere shortening. However, due to their low absorption and toxicity at higher doses, their clinical development is limited. In the present study, we examine the synergistic potential of their combination in cancer cells, which may result in a decrease in the therapeutic dosage of these compounds. We report that MST-312 and quercetin exhibit strong synergism in ovarian cancer cells with combination index range from 0.2 to 0.7. Co-treatment with MST-312 and quercetin upregulates the DNA damage and augments apoptosis when compared to treatment with either compound alone or a vehicle. We also examined the effect of these compounds on the proliferation of normal ovarian surface epithelial cells (OSEs). MST-312 has a cytoprotective impact in OSEs at lower dosages, but is inhibitory at higher doses. Quercetin did not affect the OSEs proliferation at low concentrations while at higher concentrations it is inhibitory. Notably, combination of MST-312 and quercetin had no discernible impact on OSEs. These observations have significant implications for future efforts towards maximizing efficacy in cancer therapeutics as this co-treatment specifically affects cancer cells and reduces the effective dosage of both the compounds.
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
MST-312, ≥98% (HPLC)