MilliporeSigma
  • A small molecule inhibitor of PTP1B and PTPN2 enhances T cell anti-tumor immunity.
A small molecule inhibitor of PTP1B and PTPN2 enhances T cell anti-tumor immunity.
Nature communications (2023-07-28)
Shuwei Liang, Eric Tran, Xin Du, Jiajun Dong, Harrison Sudholz, Hao Chen, Zihan Qu, Nicholas D Huntington, Jeffrey J Babon, Nadia J Kershaw, Zhong-Yin Zhang, Jonathan B Baell, Florian Wiede, Tony Tiganis
ABSTRACT
The inhibition of protein tyrosine phosphatases 1B (PTP1B) and N2 (PTPN2) has emerged as an exciting approach for bolstering T cell anti-tumor immunity. ABBV-CLS-484 is a PTP1B/PTPN2 inhibitor in clinical trials for solid tumors. Here we have explored the therapeutic potential of a related small-molecule-inhibitor, Compound-182. We demonstrate that Compound-182 is a highly potent and selective active site competitive inhibitor of PTP1B and PTPN2 that enhances T cell recruitment and activation and represses the growth of tumors in mice, without promoting overt immune-related toxicities. The enhanced anti-tumor immunity in immunogenic tumors can be ascribed to the inhibition of PTP1B/PTPN2 in T cells, whereas in cold tumors, Compound-182 elicited direct effects on both tumor cells and T cells. Importantly, treatment with Compound-182 rendered otherwise resistant tumors sensitive to α-PD-1 therapy. Our findings establish the potential for small molecule inhibitors of PTP1B and PTPN2 to enhance anti-tumor immunity and combat cancer.
Sigma-Aldrich
bpV(phen), ≥95% V basis
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid