MilliporeSigma
  • Oxytocin Modulates Osteogenic Commitment in Human Adipose-Derived Stem Cells.
Oxytocin Modulates Osteogenic Commitment in Human Adipose-Derived Stem Cells.
International journal of molecular sciences (2023-07-14)
Giovannamaria Petrocelli, Provvidenza Maria Abruzzo, Luca Pampanella, Riccardo Tassinari, Serena Marini, Elena Zamagni, Carlo Ventura, Federica Facchin, Silvia Canaider
ABSTRACT
Human adipose-derived stem cells (hASCs) are commonly harvested in minimally invasive contexts with few ethical concerns, and exhibit self-renewal, multi-lineage differentiation, and trophic signaling that make them attractive candidates for cell therapy approaches. The identification of natural molecules that can modulate their biological properties is a challenge for many researchers. Oxytocin (OXT) is a neurohypophyseal hormone that plays a pivotal role in the regulation of mammalian behavior, and is involved in health and well-being processes. Here, we investigated the role of OXT on hASC proliferation, migratory ability, senescence, and autophagy after a treatment of 72 h; OXT did not affect hASC proliferation and migratory ability. Moreover, we observed an increase in SA-β-galactosidase activity, probably related to the promotion of the autophagic process. In addition, the effects of OXT were evaluated on the hASC differentiation ability; OXT promoted osteogenic differentiation in a dose-dependent manner, as demonstrated by Alizarin red staining and gene/protein expression analysis, while it did not affect or reduce adipogenic differentiation. We also observed an increase in the expression of autophagy marker genes at the beginning of the osteogenic process in OXT-treated hASCs, leading us to hypothesize that OXT could promote osteogenesis in hASCs by modulating the autophagic process.
Sigma-Aldrich
Oxytocin acetate salt hydrate, ≥97% (HPLC)