MilliporeSigma
  • Enhancement of airway epithelial cell differentiation by pulmonary endothelial cell co-culture.
Enhancement of airway epithelial cell differentiation by pulmonary endothelial cell co-culture.
Stem cell research (2022-11-18)
Umida Burkhanova, Ann Harris, Shih-Hsing Leir
ABSTRACT
Cross-talk between lung epithelial cells and their microenvironment has an important physiological role in development. Using an in vitro model of differentiation of human induced pluripotent stem cells (iPSCs) to air-liquid interface (ALI)-cultured lung epithelial cells, we investigated the contribution of the microenvironment to maintenance of the lung progenitor cell state. Our protocol modeled in vivo cell-to-matrix and cell-to-cell interactions. These included growth of iPSCs on inserts coated with different basement membrane proteins (collagen I, IV, fibronectin, heparan sulfate or Matrigel plus collagen IV) and co-culture with human pulmonary microvascular endothelial cells (HPMECs). Marker gene expression was measured by RT-qPCR and protein expression and localization was confirmed by immunocytochemistry. The results showed that iPSCs grown on collagen IV had the highest success rate in terms of differentiation to robust ALI-cultured lung epithelial cells, followed by fibronectin, collagen I and heparan sulfate. Coating with Matrigel mixed with collagen IV further increased the success rate to > 97 %. Co-culture of iPSCs with HPMECs enhanced the expression of key airway lineage markers (NKX2.1, KRT5, TP63, MUC5AC, MUC16, FOXJ1, CFTR and SCGB1A1) during ALI culture. Cross-talk between iPSCs and their microenvironment during cell differentiation had a significant effect on lung epithelial cell differentiation in these 3D in vitro models. Both matrix proteins and endothelial cells play critical roles in the differentiation of lung progenitor cells.
Sigma-Aldrich
Anti-SCGB1A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution