MilliporeSigma
  • ADAR1 masks the cancer immunotherapeutic promise of ZBP1-driven necroptosis.
ADAR1 masks the cancer immunotherapeutic promise of ZBP1-driven necroptosis.
Nature (2022-05-26)
Ting Zhang, Chaoran Yin, Aleksandr Fedorov, Liangjun Qiao, Hongliang Bao, Nazar Beknazarov, Shiyu Wang, Avishekh Gautam, Riley M Williams, Jeremy Chase Crawford, Suraj Peri, Vasily Studitsky, Amer A Beg, Paul G Thomas, Carl Walkley, Yan Xu, Maria Poptsova, Alan Herbert, Siddharth Balachandran
ABSTRACT
Only a small proportion of patients with cancer show lasting responses to immune checkpoint blockade (ICB)-based monotherapies. The RNA-editing enzyme ADAR1 is an emerging determinant of resistance to ICB therapy and prevents ICB responsiveness by repressing immunogenic double-stranded RNAs (dsRNAs), such as those arising from the dysregulated expression of endogenous retroviral elements (EREs)1-4. These dsRNAs trigger an interferon-dependent antitumour response by activating A-form dsRNA (A-RNA)-sensing proteins such as MDA-5 and PKR5. Here we show that ADAR1 also prevents the accrual of endogenous Z-form dsRNA elements (Z-RNAs), which were enriched in the 3' untranslated regions of interferon-stimulated mRNAs. Depletion or mutation of ADAR1 resulted in Z-RNA accumulation and activation of the Z-RNA sensor ZBP1, which culminated in RIPK3-mediated necroptosis. As no clinically viable ADAR1 inhibitors currently exist, we searched for a compound that can override the requirement for ADAR1 inhibition and directly activate ZBP1. We identified a small molecule, the curaxin CBL0137, which potently activates ZBP1 by triggering Z-DNA formation in cells. CBL0137 induced ZBP1-dependent necroptosis in cancer-associated fibroblasts and reversed ICB unresponsiveness in mouse models of melanoma. Collectively, these results demonstrate that ADAR1 represses endogenous Z-RNAs and identifies ZBP1-mediated necroptosis as a new determinant of tumour immunogenicity masked by ADAR1. Therapeutic activation of ZBP1-induced necroptosis provides a readily translatable avenue for rekindling the immune responsiveness of ICB-resistant human cancers.
Sigma-Aldrich
Anti-KiSS-1 Antibody, clone 8H4.1, clone 8H4.1, from mouse
Sigma-Aldrich
Anti-phospho-MLKL (Ser345) Antibody, clone 7C6.1, clone 7C6.1, from mouse