MilliporeSigma
  • An engineered Sox17 induces somatic to neural stem cell fate transitions independently from pluripotency reprogramming.
An engineered Sox17 induces somatic to neural stem cell fate transitions independently from pluripotency reprogramming.
Science advances (2023-08-23)
Mingxi Weng, Haoqing Hu, Matthew S Graus, Daisylyn Senna Tan, Ya Gao, Shimiao Ren, Derek Hoi Hang Ho, Jakob Langer, Markus Holzner, Yuhua Huang, Guang Sheng Ling, Cora Sau Wan Lai, Mathias Francois, Ralf Jauch
ABSTRACT
Advanced strategies to interconvert cell types provide promising avenues to model cellular pathologies and to develop therapies for neurological disorders. Yet, methods to directly transdifferentiate somatic cells into multipotent induced neural stem cells (iNSCs) are slow and inefficient, and it is unclear whether cells pass through a pluripotent state with full epigenetic reset. We report iNSC reprogramming from embryonic and aged mouse fibroblasts as well as from human blood using an engineered Sox17 (eSox17FNV). eSox17FNV efficiently drives iNSC reprogramming while Sox2 or Sox17 fail. eSox17FNV acquires the capacity to bind different protein partners on regulatory DNA to scan the genome more efficiently and has a more potent transactivation domain than Sox2. Lineage tracing and time-resolved transcriptomics show that emerging iNSCs do not transit through a pluripotent state. Our work distinguishes lineage from pluripotency reprogramming with the potential to generate more authentic cell models for aging-associated neurodegenerative diseases.
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥96% (HPLC), powder
Sigma-Aldrich
trans-2-Phenylcyclopropylamine hydrochloride