MilliporeSigma
  • Reduction of nemo-like kinase increases lysosome biogenesis and ameliorates TDP-43-related neurodegeneration.
Reduction of nemo-like kinase increases lysosome biogenesis and ameliorates TDP-43-related neurodegeneration.
The Journal of clinical investigation (2023-06-29)
Leon Tejwani, Youngseob Jung, Hiroshi Kokubu, Sowmithra Sowmithra, Luhan Ni, Changwoo Lee, Benjamin Sanders, Paul J Lee, Yangfei Xiang, Kimberly Luttik, Armand Soriano, Jennifer Yoon, Junhyun Park, Hannah H Ro, Hyoungseok Ju, Clara Liao, Sofia Massaro Tieze, Frank Rigo, Paymaan Jafar-Nejad, Janghoo Lim
ABSTRACT
Protein aggregation is a hallmark of many neurodegenerative disorders, including amyotrophic lateral sclerosis (ALS). Although mutations in TARDBP, encoding transactive response DNA-binding protein 43 kDa (TDP-43), account for less than 1% of all ALS cases, TDP-43-positive aggregates are present in nearly all ALS patients, including patients with sporadic ALS (sALS) or carrying other familial ALS-causing (fALS-causing) mutations. Interestingly, TDP-43 inclusions are also present in subsets of patients with frontotemporal dementia, Alzheimer's disease, and Parkinson's disease; therefore, methods of activating intracellular protein quality control machinery capable of clearing toxic cytoplasmic TDP-43 species may alleviate disease-related phenotypes. Here, we identify a function of nemo-like kinase (Nlk) as a negative regulator of lysosome biogenesis. Genetic or pharmacological reduction of Nlk increased lysosome formation and improved clearance of aggregated TDP-43. Furthermore, Nlk reduction ameliorated pathological, behavioral, and life span deficits in 2 distinct mouse models of TDP-43 proteinopathy. Because many toxic proteins can be cleared through the autophagy/lysosome pathway, targeted reduction of Nlk represents a potential approach to therapy development for multiple neurodegenerative disorders.
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone hVIN-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Histone H3 Antibody, CT, pan, clone A3S, rabbit monoclonal, clone A3S, Upstate®, from rabbit
Sigma-Aldrich
Anti-phospho TFEB (Ser142), from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat