MilliporeSigma
  • Efficacy and specificity of inhibitors of BCL-2 family protein interactions assessed by affinity measurements in live cells.
Efficacy and specificity of inhibitors of BCL-2 family protein interactions assessed by affinity measurements in live cells.
Science advances (2022-04-21)
Elizabeth J Osterlund, Nehad Hirmiz, James M Pemberton, Adrien Nougarède, Qian Liu, Brian Leber, Qiyin Fang, David W Andrews
ABSTRACT
Cytoplasmic and membrane-bound BCL-2 family proteins regulate apoptosis, a form of programmed cell death, via dozens of binary protein interactions confounding measurement of the effects of inhibitors in live cells. In cancer, apoptosis is frequently dysregulated, and cell survival depends on antiapoptotic proteins binding to and inhibiting proapoptotic BH3 proteins. The clinical success of BH3 mimetic inhibitors of antiapoptotic proteins has spawned major efforts by the pharmaceutical industry to develop molecules with different specificities and higher affinities. Here, quantitative fast fluorescence lifetime imaging microscopy enabled comparison of BH3 mimetic drugs in trials and preclinical development by measuring drug effects on binding affinities of interacting protein pairs in live cells. Both selectivity and efficacy were assessed for 15 inhibitors of four antiapoptotic proteins for each of six BH3 protein ligands. While many drugs target the designed interaction, most also have unexpected selectivity and poor efficacy in cells.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Pepstatin A, microbial, ≥90% (HPLC)
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized, ~80% (HPCE), crystalline (fine), white, ≥3500 U/mg
Sigma-Aldrich
Fetal Bovine Serum, Canada origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Chymostatin, microbial