MilliporeSigma
  • TAK1 Improves Cognitive Function via Suppressing RIPK1-Driven Neuronal Apoptosis and Necroptosis in Rats with Chronic Hypertension.
TAK1 Improves Cognitive Function via Suppressing RIPK1-Driven Neuronal Apoptosis and Necroptosis in Rats with Chronic Hypertension.
Aging and disease (2023-05-17)
Jing Yang, Pei Sun, Xiangming Xu, Xiaolu Liu, Linfang Lan, Ming Yi, Chi Xiao, Ruichen Ni, Yuhua Fan
ABSTRACT
Chronic hypertension is a major risk factor for cognitive impairment, which can promote neuroinflammation and neuronal loss in the central nervous system. Transforming growth factor β-activated kinase 1 (TAK1) is a key molecular component in determining cell fate and can be activated by inflammatory cytokines. This study aimed to investigate the role of TAK1 in mediating neuronal survival in the cerebral cortex and hippocampus under chronic hypertensive conditions. To that end, we used stroke-prone renovascular hypertension rats (RHRSP) as chronic hypertension models. Adeno-associated virus (AAV) designed to overexpress or knock down TAK1 expression were injected into the lateral ventricles of rats and the subsequent effects on cognitive function and neuronal survival under chronic hypertensive conditions were assessed. We found that, TAK1 knockdown in RHRSP markedly increased neuronal apoptosis and necroptosis and induced cognitive impairment, which could be reversed by Nec-1s, an inhibitor of receptor interacting protein kinase 1 (RIPK1). In contrast, overexpression of TAK1 in RHRSP significantly suppressed neuronal apoptosis and necroptosis and improved cognitive function. Further knockdown of TAK1 in sham-operated rats received similar phenotype with RHRSP. The results have been verified in vitro. In this study, we provide in vivo and in vitro evidence that TAK1 improves cognitive function by suppressing RIPK1-driven neuronal apoptosis and necroptosis in rats with chronic hypertension.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, Alexa Fluor 488, clone GA5, Chemicon®, from mouse