MilliporeSigma
  • Microglial CD68 and L-ferritin upregulation in response to phosphorylated-TDP-43 pathology in the amyotrophic lateral sclerosis brain.
Microglial CD68 and L-ferritin upregulation in response to phosphorylated-TDP-43 pathology in the amyotrophic lateral sclerosis brain.
Acta neuropathologica communications (2023-04-29)
Molly E V Swanson, Miran Mrkela, Helen C Murray, Maize C Cao, Clinton Turner, Maurice A Curtis, Richard L M Faull, Adam K Walker, Emma L Scotter
ABSTRACT
Microglia, the innate immune cells of the brain, are activated by damage or disease. In mouse models of amyotrophic lateral sclerosis (ALS), microglia shift from neurotrophic to neurotoxic states with disease progression. It remains unclear how human microglia change relative to the TAR DNA-binding protein 43 (TDP-43) aggregation that occurs in 97% of ALS cases. Here we examine spatial relationships between microglial activation and TDP-43 pathology in brain tissue from people with ALS and from a TDP-43-driven ALS mouse model. Post-mortem human brain tissue from the Neurological Foundation Human Brain Bank was obtained from 10 control and 10 ALS cases in parallel with brain tissue from a bigenic NEFH-tTA/tetO-hTDP-43∆NLS (rNLS) mouse model of ALS at disease onset, early disease, and late disease stages. The spatiotemporal relationship between microglial activation and ALS pathology was determined by investigating microglial functional marker expression in brain regions with low and high TDP-43 burden at end-stage human disease: hippocampus and motor cortex, respectively. Sections were immunohistochemically labelled with a two-round multiplexed antibody panel against; microglial functional markers (L-ferritin, HLA-DR, CD74, CD68, and Iba1), a neuronal marker, an astrocyte marker, and pathological phosphorylated TDP-43 (pTDP-43). Single-cell levels of microglial functional markers were quantified using custom analysis pipelines and mapped to anatomical regions and ALS pathology. We identified a significant increase in microglial Iba1 and CD68 expression in the human ALS motor cortex, with microglial CD68 being significantly correlated with pTDP-43 pathology load. We also identified two subpopulations of microglia enriched in the ALS motor cortex that were defined by high L-ferritin expression. A similar pattern of microglial changes was observed in the rNLS mouse, with an increase first in CD68 and then in L-ferritin expression, with both occurring only after pTDP-43 inclusions were detectable. Our data strongly suggest that microglia are phagocytic at early-stage ALS but transition to a dysfunctional state at end-stage disease, and that these functional states are driven by pTDP-43 aggregation. Overall, these findings enhance our understanding of microglial phenotypes and function in ALS.
Sigma-Aldrich
Lectin from Ulex europaeus (gorse, furze), biotin conjugate, lyophilized powder
Sigma-Aldrich
Anti-NeuN Antibody, serum, from guinea pig