MilliporeSigma
  • Combination Ad26.RSV.preF/preF protein vaccine induces superior protective immunity compared with individual vaccine components in preclinical models.
Combination Ad26.RSV.preF/preF protein vaccine induces superior protective immunity compared with individual vaccine components in preclinical models.
NPJ vaccines (2023-03-24)
Eirikur Saeland, Leslie van der Fits, Renske Bolder, Marjolein Heemskerk-van der Meer, Joke Drijver, Yolinda van Polanen, Cornelis Vaneman, Lisanne Tettero, Freek Cox, Jan Serroyen, Matthew J Jorgensen, Johannes P M Langedijk, Hanneke Schuitemaker, Benoit Callendret, Roland C Zahn
ABSTRACT
Respiratory syncytial virus (RSV) is a leading cause of severe respiratory disease for which no licensed vaccine is available. We have previously shown that a prefusion (preF) conformation-stabilized RSV F protein antigen and an adenoviral vector encoding RSV preF protein (Ad26.RSV.preF) are immunogenic and protective in animals when administered as single components. Here, we evaluated a combination of the 2 components, administered as a single injection. Strong induction of both humoral and cellular responses was shown in RSV-naïve and pre-exposed mice and pre-exposed African green monkeys (AGMs). Both components of the combination vaccine contributed to humoral immune responses, while the Ad26.RSV.preF component was the main contributor to cellular immune responses in both mice and AGMs. Immunization with the combination elicited superior protection against RSV A2 challenge in cotton rats. These results demonstrate the advantage of a combination vaccine and support further clinical development.