MilliporeSigma
  • TREM2 Agonism with a Monoclonal Antibody Attenuates Tau Pathology and Neurodegeneration.
TREM2 Agonism with a Monoclonal Antibody Attenuates Tau Pathology and Neurodegeneration.
Cells (2023-06-10)
Michael Fassler, Clara Benaim, Jacob George
ABSTRACT
TREM2 is a membrane receptor expressed on microglia that plays a pivotal role in the organization and function of these innate immune cell components within the neurodegenerated brain. Whereas TREM2 deletion has been studied extensively in experimental beta-amyloid and Tau-based models of Alzheimer's disease, its engagement, and subsequent agonism have not been tested in the context of Tau pathology. Herein, we explored the effects of Ab-T1, an agonistic TREM2 monoclonal antibody on Tau uptake, phosphorylation, seeding, and spreading as well as its therapeutic efficacy in a Tauopathy model. Ab-T1 enhanced the uptake of misfolded Tau to microglia and induced a non-cell autonomous attenuation of spontaneous Tau seeding and phosphorylation in primary neurons from human Tau transgenic mice. Ex vivo, incubation with Ab-T1 led to a significant reduction in the seeding of Tau pathology in the hTau murine organoid brain system. Systemic administration of Ab-T1 resulted in reduced Tau pathology and propagation when hTau was stereotactically injected into the hemispheres of hTau mice. Intraperitoneal treatment with Ab-T1 lead to attenuation of cognitive decline in the hTau mice that was associated with reduced neurodegeneration and synaptic preservation with amelioration of the global neuroinflammatory program. Collectively, these observations show that TREM2 engagement with an agonistic antibody result in reduced Tau burden concomitant with attenuated neurodegeneration ascribed to the education of resident microglia. These results may suggest that despite the opposing results with regard to the effect of TREM2 knockout in experimental Tau-based model systems, receptor engagement and activation by Ab-T1 appears to possess beneficial effects with respect to the various mechanisms mediating Tau-driven neurodegeneration.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Anti-Tau Antibody, clone T49 (Not human), clone T49, from mouse
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse