MilliporeSigma
  • Pulmonary neuroendocrine cells sense succinate to stimulate myoepithelial cell contraction.
Pulmonary neuroendocrine cells sense succinate to stimulate myoepithelial cell contraction.
Developmental cell (2022-09-16)
Wenjie Yu, Thomas O Moninger, Michael V Rector, David A Stoltz, Michael J Welsh
ABSTRACT
Pulmonary neuroendocrine cells (PNECs) are rare airway cells with potential sensory capacity linked to vagal neurons and immune cells. How PNECs sense and respond to external stimuli remains poorly understood. We discovered PNECs located within pig and human submucosal glands, a tissue that produces much of the mucus that defends the lung. These PNECs sense succinate, an inflammatory molecule in liquid lining the airway surface. The results indicate that succinate migrates down the submucosal gland duct to the acinus, where it triggers apical succinate receptors, causing PNECs to release ATP. The short-range ATP signal stimulates the contraction of myoepithelial cells wrapped tightly around the submucosal glands. Succinate-triggered gland contraction may complement the action of neurotransmitters that induce mucus release but not gland contraction to promote mucus ejection onto the airway surface. These findings identify a local circuit in which rare PNECs within submucosal glands sense an environmental cue to orchestrate the function of airway glands.
Sigma-Aldrich
Anti-Vesicular Nucleotide Transporter (VNUT Antibody), serum, from guinea pig
Sigma-Aldrich
Anti-MUC5B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Substance P, Neuropeptide found in high concentrations in the gut.