MilliporeSigma
  • Evidence for Phosphorylation-Dependent, Dynamic, Regulation of mGlu5 and Homer2 in Expression of Cocaine Aversion in Mice.
Evidence for Phosphorylation-Dependent, Dynamic, Regulation of mGlu5 and Homer2 in Expression of Cocaine Aversion in Mice.
eNeuro (2023-03-28)
Karen K Szumlinski, Jacqueline Beltran, Eliyana van Doren, C Leonardo Jimenez Chavez, Racquel D Domingo-Gonzalez, Cindy M Reyes, Alexis W Ary, Andrew Lang, Weiruo Guo, Paul F Worley, Kimberly M Huber
ABSTRACT
Cocaine-induced changes in the expression of the glutamate-related scaffolding protein Homer2 influence this drug's psychostimulant and rewarding properties. In response to neuronal activity, Homer2 is phosphorylated on S117/S216 by calcium-calmodulin kinase IIα (CaMKIIα), which induces a rapid dissociation of mGlu5-Homer2 scaffolds. Herein, we examined the requirement for Homer2 phosphorylation in cocaine-induced changes in mGlu5-Homer2 coupling, to include behavioral sensitivity to cocaine. For this, mice with alanine point mutations at (S117/216)-Homer2 (Homer2AA/AA ) were generated, and we determined their affective, cognitive and sensorimotor phenotypes, as well as cocaine-induced changes in conditioned reward and motor hyperactivity. The Homer2AA/AA mutation prevented activity-dependent phosphorylation of S216 Homer2 in cortical neurons, but Homer2AA/AA mice did not differ from wild-type (WT) controls with respect to Morris maze performance, acoustic startle, spontaneous or cocaine-induced locomotion. Homer2AA/AA mice exhibited signs of hypoanxiety similar to the phenotype of transgenic mice with a deficit in signal-regulated mGluR5 phosphorylation (Grm5AA/AA ). However, opposite of Grm5AA/AA mice, Homer2AA/AA mice were less sensitive to the aversive properties of high-dose cocaine under both place-conditioning and taste-conditioning procedures. Acute injection with cocaine caused dissociation of mGluR5 and Homer2 in striatal lysates from WT, but not Homer2AA/AA mice, suggesting a molecular basis for the deficit in cocaine aversion. These findings indicate that CaMKIIα-dependent phosphorylation of Homer2 gates the negative motivational valence of high-dose cocaine via regulation of mGlu5 binding, furthering an important role for dynamic changes in mGlu5-Homer interactions in addiction vulnerability.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-phospho-CaMK2 (pThr286) antibody produced in rabbit, affinity isolated antibody