MilliporeSigma
  • Osteoclast-mediated resorption primes the skeleton for successful integration during axolotl limb regeneration.
Osteoclast-mediated resorption primes the skeleton for successful integration during axolotl limb regeneration.
eLife (2022-10-12)
Camilo Riquelme-Guzmán, Stephanie L Tsai, Karen Carreon Paz, Congtin Nguyen, David Oriola, Maritta Schuez, Jan Brugués, Joshua D Currie, Tatiana Sandoval-Guzmán
ABSTRACT
Early events during axolotl limb regeneration include an immune response and the formation of a wound epithelium. These events are linked to a clearance of damaged tissue prior to blastema formation and regeneration of the missing structures. Here, we report the resorption of calcified skeletal tissue as an active, cell-driven, and highly regulated event. This process, carried out by osteoclasts, is essential for a successful integration of the newly formed skeleton. Indeed, the extent of resorption is directly correlated with the integration efficiency, and treatment with zoledronic acid resulted in osteoclast function inhibition and failed tissue integration. Moreover, we identified the wound epithelium as a regulator of skeletal resorption, likely releasing signals involved in recruitment/differentiation of osteoclasts. Finally, we reported a correlation between resorption and blastema formation, particularly, a coordination of resorption with cartilage condensation. In sum, our results identify resorption as a major event upon amputation, playing a critical role in the overall process of skeletal regeneration.
Product Description
Roche
SP6 RNA Polymerase, from Escherichia coli BL 21/pSR3
Roche
T7 RNA Polymerase, from Escherichia coli BL 21/pAR 1219
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep