MilliporeSigma
  • Culture of Escherichia coli in SOC medium improves the cloning efficiency of toxic protein genes.
Culture of Escherichia coli in SOC medium improves the cloning efficiency of toxic protein genes.
Analytical biochemistry (2009-07-23)
Qiao-Yang Sun, Ling-Wen Ding, Liang-Liang He, Yong-Bin Sun, Jun-Li Shao, Ming Luo, Zeng-Fu Xu
ABSTRACT
In this paper, we report a useful protocol for cloning toxic protein genes. Use of the SOC medium, which is a glucose-containing rich medium, significantly improved the transformation efficiency of a recombinant plasmid containing a toxic plant subtilase SaSBT1 cDNA. Both glucose and rich nutrients present in the SOC medium prevented the unintended activation of the lac promoter carried on the cloning vector, and led to significantly improved transformation efficiency of recombinant plasmids containing toxic protein genes and an increased rate of transformant growth.
Sigma-Aldrich
SOC Medium, For use in transformation
Pricing and availability is not currently available.