MilliporeSigma
  • Crystal violet-based assay for the assessment of bacterial biofilm formation in medical tubing.
Crystal violet-based assay for the assessment of bacterial biofilm formation in medical tubing.
Journal of microbiological methods (2022-12-17)
Marcin Borowicz, Dorota M Krzyżanowska, Sylwia Jafra
ABSTRACT
Crystal violet-based assay is widely used to evaluate the early stages of bacterial biofilm formation. Here, we modified it to study biofilm formation in medical tubing. While cost-effective and requiring no sophisticated equipment, our method provided quantitative and qualitative distinctions between the ability of four bacterial strains to colonize PVC catheters.
Sigma-Aldrich
LB Broth, Miller - Novagen, Novagen®, granular, pack of 500 g or 5 kg, for Escherichia coli